Merck
  • Evaluation of the efficacy of a recombinant subunit West Nile vaccine in Syrian golden hamsters.
Evaluation of the efficacy of a recombinant subunit West Nile vaccine in Syrian golden hamsters.
The American journal of tropical medicine and hygiene (2008-12-05)
Marina T Siirin, Amelia P A Travassos da Rosa, Patrick Newman, Carolyn Weeks-Levy, Beth-Ann Coller, Shu-Yuan Xiao, Michael M Lieberman, Douglas M Watts
ABSTRACT
The efficacy of a recombinant subunit West Nile (WN) vaccine candidate was determined in a hamster model of encephalitis. Animals included young, aged, and immunocompromised animals in an effort to simulate key groups at risk of WN virus-induced disease. Groups of aged (12 month old), weanling, and adult hamsters rendered leukopenic after immunization were immunized subcutaneously with a WN virus recombinant envelope protein (WN-80E) with or without WN virus non-structural protein 1 (NS1) mixed with adjuvant or adjuvant alone. A challenge dose of wild-type WN virus was administered to produce 40-100% mortality in the control hamsters. The recombinant antigen preparations containing WN-80E with or without WN NS1 gave similar results. Hamsters in both groups had a strong antibody response after immunization, and none of the aged or weanling animals became ill or developed detectable viremia after challenge with WN virus at 2 weeks after booster vaccination. However, mortality among the control animals (administered adjuvant without antigen) at 2 weeks after booster challenge was 40-60%. In hamsters rendered leukopenic after immunization, survival rates up to 80% were observed, and a low-level viremia was detected in the vaccinated and challenged hamsters. The survival rate was significantly (P<0.05) higher in animals vaccinated with a higher dose of WN-80E than a lower dose. The addition of NS1 did not significantly affect survival after challenge. In contrast, all of the control animals that received adjuvant only developed a very high level of viremia, and the mortality rate was 100%. These findings indicate that the recombinant WN vaccines induced antibody in and afforded protection to young and aged hamsters and immunosuppressed hamsters.
Sigma-Aldrich
Neutral Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Neutral Red, Dye content ≥90 %
Sigma-Aldrich
Neutral Red, for microscopy (Bact., Bot., Fl., Hist., Vit.), indicator (pH 6.8-8.0)
Sigma-Aldrich
Neutral Red, certified by the BSC
Sigma-Aldrich
Guinea pig serum