Merck
  • Inhibition of DNMT3B expression in activated hepatic stellate cells overcomes chemoresistance in the tumor microenvironment of hepatocellular carcinoma.
Inhibition of DNMT3B expression in activated hepatic stellate cells overcomes chemoresistance in the tumor microenvironment of hepatocellular carcinoma.
Scientific reports (2024-01-04)
Yeonhwa Song, Namjeong Kim, Jinyeong Heo, David Shum, Taemoo Heo, Haeng Ran Seo
ABSTRACT
Hepatocellular carcinoma (HCC) is a complex disease associated with a plethora of environmental and genetic/hereditary causative risk factors, more so than other oncological indications. Additionally, patients with HCC exhibit fibrosis, cirrhosis, and liver-related disease. This complicated etiology can affect the disease course and likely contributes to its poor prognosis. In this study, we aimed to improve HCC therapy by evaluating combination treatment using anti-cancer and anti-fibrosis drugs via identification of novel anti-fibrosis drugs. We performed high-throughput screening of 10,000 compounds to identify hepatic fibrosis inhibitors through morphometry analysis of multicellular hepatic spheroid (MCHS) models and identified CHIR-99021 as a candidate anti-fibrotic drug. Treatment with CHIR-99021 induced loss of cell-cell interactions and suppression of extracellular matrix-related protein expression via reprogramming of hepatic stellate cell (HSC) activation in MCHSs. In particular, CHIR-99021 regulated DNMT3B expression only in activated HSCs. Moreover, CHIR-99021 markedly improved the efficacy of sorafenib in HCC- multicellular tumor spheroids in vitro and through induction of apoptosis by decreasing DNMT3B expression in vivo. In summary, these findings suggest that targeting HSC reprogramming by attenuation of DNMT3B expression in the tumor environment might represent a promising therapeutic strategy for liver fibrosis and HCC.
Sigma-Aldrich
SB 216763, >98% (HPLC)
Sigma-Aldrich
SB 415286, ≥98% (HPLC)