Merck
  • Progressive tumor features accompany epithelial-mesenchymal transition induced in mitochondrial DNA-depleted cells.
Progressive tumor features accompany epithelial-mesenchymal transition induced in mitochondrial DNA-depleted cells.
Cancer science (2008-08-30)
Akihiro Naito, Cody C Cook, Takatsugu Mizumachi, Mian Wang, Cheng-Hui Xie, Teresa T Evans, Thomas Kelly, Masahiro Higuchi
ABSTRACT
The growth of LNCaP, a human prostate adenocarcinoma cell line, and MCF-7, a human breast adenocarcinoma cell line, is initially hormone dependent. We previously demonstrated that LNrho0-8 and MCFrho0, derived from LNCaP and MCF-7 by depleting mitochondrial DNA (mtDNA), exhibited hormone-independent growth that led to progressed phenotypes. Here, we demonstrate that LNrho0-8 and MCFrho0 have invasive characters as evaluated by the ability of invasion through the extracellular matrix (ECM) in vitro. In addition, the induction of vimentin and the repression of E-cadherin expression in rho0 cells indicate that they are mesenchymal cells. Since LNrho0-8 and MCFrho0 were derived from epithelial cancer cell lines, LNCaP and MCF-7 must have lost epithelial features and gained the mesenchymal phenotype by epithelial-mesenchymal transition (EMT) during the mtDNA depletion. In the rho0 cell lines, the Raf/MAPK signaling cascade was highly activated together with the expressions of transforming growth factor-beta (TGF-beta) and type I TGF-beta receptor (TGF-betaRI). EMT requires cooperation of TGF-beta signaling with activation of the Raf/MAPK cascade, suggesting that EMT was induced in mtDNA depleted cells resulting in the acquisition of progressive tumor features, such as higher invasiveness and loss of hormone dependent growth. Our results indicate that decreasing mtDNA content induces EMT, enabling the progressive phenotypes observed in cancer.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, For use as a marker in SDS-PAGE
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥97% (agarose gel electrophoresis), 1X Crystallized
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Essentially fatty acid free
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Insulin from bovine pancreas, γ-irradiated, BioXtra, suitable for cell culture, potency: ≥20 units/mg (USP units), lyophilized powder
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, chromatographically purified, New Zealand origin, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, low endotoxin, BioReagent, suitable for cell culture, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)