Merck
  • Knockout of PA200 improves proteasomal degradation and myelination in a proteotoxic neuropathy.
Knockout of PA200 improves proteasomal degradation and myelination in a proteotoxic neuropathy.
Life science alliance (2024-02-10)
Jordan Js VerPlank, Joseph M Gawron, Nicholas J Silvestri, Lawrence Wrabetz, Maria Laura Feltri
ABSTRACT
The cellular response to a decrease in protein degradation by 26S proteasomes in chronic diseases is poorly understood. Pharmacological inhibition of proteasomes increases the expression of proteasome subunits and Proteasome Activator 200 (PA200), an alternative proteasome activator. In the S63del mouse model of the peripheral neuropathy Charcot Marie Tooth 1B (CMT1B), proteasomal protein degradation is decreased and proteasome gene expression is increased. Here, we show an increase in PA200 and PA200-bound proteasomes in the peripheral nerves of S63del mice. To test genetically whether the upregulation of PA200 was compensatory, we generated S63del//PA200-/- mice. Unexpectedly, in the sciatic nerves of these mice, there was greater proteasomal protein degradation than in S63del, less polyubiquitinated proteins and markers of the unfolded protein response, and a greater amount of assembled, active 26S proteasomes. These changes were not seen in PA200-/- controls and were therefore specific to the neuropathy. Furthermore, in S63del//PA200-/- mice, myelin thickness and nerve conduction were restored to WT levels. Thus, the upregulation of PA200 is maladaptive in S63del mice and its genetic ablation prevented neuropathy.
Sigma-Aldrich
Anti-PMP22 antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.