Merck
  • Impact of the RNA chaperone Hfq on the fitness and virulence potential of uropathogenic Escherichia coli.
Impact of the RNA chaperone Hfq on the fitness and virulence potential of uropathogenic Escherichia coli.
Infection and immunity (2008-05-07)
Richard R Kulesus, Karen Diaz-Perez, E Susan Slechta, Danelle S Eto, Matthew A Mulvey
ABSTRACT
Hfq is a bacterial RNA chaperone involved in the posttranscriptional regulation of many stress-inducible genes via small noncoding RNAs. Here, we show that Hfq is critical for the uropathogenic Escherichia coli (UPEC) isolate UTI89 to effectively colonize the bladder and kidneys in a murine urinary tract infection model system. The disruption of hfq did not affect bacterial adherence to or invasion of host cells but did limit the development of intracellular microcolonies by UTI89 within the terminally differentiated epithelial cells that line the lumen of the bladder. In vitro, the hfq mutant was significantly impaired in its abilities to handle the antibacterial cationic peptide polymyxin B and reactive nitrogen and oxygen radicals and to grow in acidic medium (pH 5.0). Relative to the wild-type strain, the hfq mutant also had a substantially reduced migration rate on motility agar and was less prone to form biofilms. Hfq activities are known to impact the regulation of both the stationary-phase sigma factor RpoS (sigma(S)) and the envelope stress response sigma factor RpoE (sigma(E)). Although we saw similarities among hfq, rpoS, and rpoE deletion mutants in our assays, the rpoE and hfq mutants were phenotypically the most alike. Cumulatively, our data indicate that Hfq likely affects UPEC virulence-related phenotypes primarily by modulating membrane homeostasis and envelope stress response pathways.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dimethyl sulfoxide, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium nitrite, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%