Merck
  • Leucine Supplementation Improves Diastolic Function in HFpEF by HDAC4 Inhibition.
Leucine Supplementation Improves Diastolic Function in HFpEF by HDAC4 Inhibition.
Cells (2023-11-10)
Paula Ketilly Nascimento Alves, Antje Schauer, Antje Augstein, Anita Männel, Peggy Barthel, Dirk Joachim, Janet Friedrich, Maria-Elisa Prieto, Anselmo Sigari Moriscot, Axel Linke, Volker Adams
ABSTRACT
Heart failure with preserved ejection fraction (HFpEF) is a complex syndrome associated with a high morbidity and mortality rate. Leucine supplementation has been demonstrated to attenuate cardiac dysfunction in animal models of cachexia and heart failure with reduced ejection fraction (HFrEF). So far, no data exist on leucine supplementation on cardiac function in HFpEF. Thus, the current study aimed to investigate the effect of leucine supplementation on myocardial function and key signaling pathways in an established HFpEF rat model. Female ZSF1 rats were randomized into three groups: Control (untreated lean rats), HFpEF (untreated obese rats), and HFpEF_Leu (obese rats receiving standard chow enriched with 3% leucine). Leucine supplementation started at 20 weeks of age after an established HFpEF was confirmed in obese rats. In all animals, cardiac function was assessed by echocardiography at baseline and throughout the experiment. At the age of 32 weeks, hemodynamics were measured invasively, and myocardial tissue was collected for assessment of mitochondrial function and for histological and molecular analyses. Leucine had already improved diastolic function after 4 weeks of treatment. This was accompanied by improved hemodynamics and reduced stiffness, as well as by reduced left ventricular fibrosis and hypertrophy. Cardiac mitochondrial respiratory function was improved by leucine without alteration of the cardiac mitochondrial content. Lastly, leucine supplementation suppressed the expression and nuclear localization of HDAC4 and was associated with Protein kinase A activation. Our data show that leucine supplementation improves diastolic function and decreases remodeling processes in a rat model of HFpEF. Beneficial effects were associated with HDAC4/TGF-β1/Collagenase downregulation and indicate a potential use in the treatment of HFpEF.
Sigma-Aldrich
Anti-Collagen Antibody, Type IV, Chemicon®, from rabbit
Sigma-Aldrich
Anti-phospho-PKA CAT (pThr197) antibody produced in rabbit, affinity isolated antibody