Merck
  • Knockdown of BAP31 Downregulates Galectin-3 to Inhibit the Wnt/β-Catenin Signaling Pathway to Modulate 5-FU Chemosensitivity and Cancer Stemness in Colorectal Cancer.
Knockdown of BAP31 Downregulates Galectin-3 to Inhibit the Wnt/β-Catenin Signaling Pathway to Modulate 5-FU Chemosensitivity and Cancer Stemness in Colorectal Cancer.
International journal of molecular sciences (2023-09-28)
Jingjing Liu, Qi Zhang, Jiyu Wang, Changli Wang, Tian Lan, Tianyi Wang, Bing Wang
ABSTRACT
Increased stemness is causally linked to the development of chemoresistance in cancers. B-cell receptor-associated protein 31 (BAP31) has been identified to play an oncogenic role in many types of cancer. However, the role of BAP31 in 5-fluorouracil (5-FU) chemosensitivity and stemness of colorectal cancer (CRC) is still unknown. The aim of this study was to investigate the biological function and molecular mechanism of BAP31 in regulating 5-FU chemosensitivity and stemness. The correlation between BAP31 expression and 5-FU chemosensitivity was examined using 3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2-H-tetrazolium bromide and colony formation assays. Cancer stemness was analyzed using tumor sphere formation and Western blot assays. Western blot and immunofluorescence analyses of the knockdown cell lines were performed to explore the possible mechanisms. Finally, we investigated the function of BAP31 by constructing xenograft nude mouse models in vivo. In this study, we demonstrated that BAP31 was increased in CRC cells, and knockdown of BAP31 reduced the half maximal inhibitory concentration (IC50) of 5-FU, while this effect was reversed by overexpression of BAP31. In addition, knockdown of BAP31 substantially reduced the stemness of CRC cells in vitro. Consistently, knockdown of BAP31 significantly suppressed the tumorigenicity and stemness of CRC in vivo. The functional study further suggested that knockdown of BAP31 downregulated galectin-3 to inhibit the accumulation of β-catenin, which in turn repressed the transcription of downstream target genes (c-MYC, SOX2) of the Wnt/β-catenin signaling pathway. Knockdown of BAP31 reduced stemness by inhibiting the Wnt/β-catenin signaling pathway to increase 5-FU chemosensitivity. Importantly, intrabodies against BAP31 suppressed tumor growth and enhanced the antitumor effects of 5-FU in vivo. Therefore, using intrabodies against BAP31 may be a strategy for improving the antitumor effect of 5-FU in CRC.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture