Merck
  • Development of Inapparent Dengue Associated With Increased Antibody Levels to Aedes aegypti Salivary Proteins: A Longitudinal Dengue Cohort in Cambodia.
Development of Inapparent Dengue Associated With Increased Antibody Levels to Aedes aegypti Salivary Proteins: A Longitudinal Dengue Cohort in Cambodia.
The Journal of infectious diseases (2021-11-01)
Jessica E Manning, Sophana Chea, Daniel M Parker, Jennifer A Bohl, Sreyngim Lay, Allyson Mateja, Somnang Man, Sreynik Nhek, Aiyana Ponce, Sokunthea Sreng, Dara Kong, Soun Kimsan, Claudio Meneses, Michael P Fay, Seila Suon, Rekol Huy, Chanthap Lon, Rithea Leang, Fabiano Oliveira
ABSTRACT
We established the first prospective cohort to understand how infection with dengue virus is influenced by vector-specific determinants such as humoral immunity to Aedes aegypti salivary proteins. Children aged 2-9 years were enrolled in the PAGODAS (Pediatric Assessment Group of Dengue and Aedes Saliva) cohort with informed consent by their guardians. Children were followed semi-annually for antibodies to dengue and to proteins in Ae. aegypti salivary gland homogenate using enzyme-linked immunosorbent assays and dengue-specific neutralization titers. Children presented with fever at any time for dengue testing. From 13 July to 30 August 2018, we enrolled 771 children. At baseline, 22% (173/770) had evidence of neutralizing antibodies to 1 or more dengue serotypes. By April 2020, 51 children had symptomatic dengue while 148 dengue-naive children had inapparent dengue defined by neutralization assays. In a multivariate model, individuals with higher antibodies to Ae. aegypti salivary proteins were 1.5 times more likely to have dengue infection (hazard ratio [HR], 1.47 [95% confidence interval {CI}, 1.05-2.06]; P = .02), particularly individuals with inapparent dengue (HR, 1.64 [95% CI, 1.12-2.41]; P = .01). High levels of seropositivity to Ae. aegypti salivary proteins are associated with future development of dengue infection, primarily inapparent, in dengue-naive Cambodian children. NCT03534245.