Merck
  • Methods to Study Inflammasome Activation in the Central Nervous System: Immunoblotting and Immunohistochemistry.
Methods to Study Inflammasome Activation in the Central Nervous System: Immunoblotting and Immunohistochemistry.
Methods in molecular biology (Clifton, N.J.) (2023-08-14)
Brianna Cyr, Juan Pablo de Rivero Vaccari
ABSTRACT
The inflammasome is a multiprotein complex that is responsible for mounting an innate immune response through the activation of caspase-1 and the cleavage of interleukin-1β. This multiprotein complex plays an important role in a variety of central nervous system (CNS) diseases and conditions such as Alzheimer's disease, Parkinson's disease, multiple sclerosis, stroke, and traumatic brain injury, among others. Here we describe methodological procedures to carry out immunoblotting and immunohistochemical techniques used to study inflammasome signaling in CNS tissues (brain and spinal cord).
Sigma-Aldrich
Anti-Integrin αM [CD11b] Antibody, clone OX-42, clone OX-42, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Pyrin, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Aldh1L1 Antibody, clone N103/39, clone N103/39, from mouse
Sigma-Aldrich
Anti-O1 Antibody, clone 59, clone 59, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse