Merck
  • BC-11 is a covalent TMPRSS2 fragment inhibitor that impedes SARS-CoV-2 host cell entry.
BC-11 is a covalent TMPRSS2 fragment inhibitor that impedes SARS-CoV-2 host cell entry.
Archiv der Pharmazie (2022-11-01)
Aurélien F A Moumbock, Hoai T T Tran, Evelyn Lamy, Stefan Günther
ABSTRACT
Host cell entry of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is facilitated via priming of its spike glycoprotein by the human transmembrane protease serine 2 (TMPRSS2). Although camostat and nafamostat are two highly potent covalent TMPRSS2 inhibitors, they nevertheless did not hold promise in COVID-19 clinical trials, presumably due to their short plasma half-lives. Herein, we report an integrative chemogenomics approach based on computational modeling and in vitro enzymatic assays, for repurposing serine-targeted covalent inhibitors. This led to the identification of BC-11 as a covalent TMPRSS2 inhibitor displaying a unique selectivity profile for serine proteases, ascribable to its boronic acid warhead. BC-11 showed modest inhibition of SARS-CoV-2 (omicron variant) spike pseudotyped particles in a cell-based entry assay, and a combination of BC-11 and AHN 1-055 (a spike glycoprotein inhibitor) demonstrated better viral entry inhibition than either compound alone. Given its low molecular weight and good activity against TMPRSS2, BC-11 qualifies as a good starting point for further structural optimizations.
Sigma-Aldrich
Z-Pro-prolinal, ≥98% (HPLC)