Merck
  • Limitations of Tamoxifen Application for In Vivo Genome Editing Using Cre/ERT2 System.
Limitations of Tamoxifen Application for In Vivo Genome Editing Using Cre/ERT2 System.
International journal of molecular sciences (2022-11-27)
Leonid A Ilchuk, Nina I Stavskaya, Ekaterina A Varlamova, Alvina I Khamidullina, Victor V Tatarskiy, Vladislav A Mogila, Ksenia B Kolbutova, Sergey A Bogdan, Alexey M Sheremetov, Alexandr N Baulin, Irina A Filatova, Yulia Yu Silaeva, Maxim A Filatov, Alexandra V Bruter
ABSTRACT
Inducible Cre-dependent systems are frequently used to produce both conditional knockouts and transgenic mice with regulated expression of the gene of interest. Induction can be achieved by doxycycline-dependent transcription of the wild type gene or OH-tamoxifen-dependent nuclear translocation of the chimeric Cre/ERT2 protein. However, both of these activation strategies have some limitations. We analyzed the efficiency of knockout in different tissues and found out that it correlates with the concentration of the hydroxytamoxifen and endoxifen-the active metabolites of tamoxifen-measured by LC-MS in these tissues. We also describe two cases of Cdk8floxed/floxed/Rosa-Cre-ERT2 mice tamoxifen-induced knockout limitations. In the first case, the standard scheme of tamoxifen administration does not lead to complete knockout formation in the brain or in the uterus. Tamoxifen metabolite measurements in multiple tissues were performed and it has been shown that low recombinase activity in the brain is due to the low levels of tamoxifen active metabolites. Increase of tamoxifen dosage (1.5 fold) and duration of activation (from 5 to 7 days) allowed us to significantly improve the knockout rate in the brain, but not in the uterus. In the second case, knockout induction during embryonic development was impossible due to the negative effect of tamoxifen on gestation. Although DNA editing in the embryos was achieved in some cases, the treatment led to different complications of the pregnancy in wild-type female mice. We propose to use doxycycline-induced Cre systems in such models.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
(Z)-Endoxifen, ≥98% (HPLC)