Merck
  • Linear ubiquitination induces NEMO phase separation to activate NF-κB signaling.
Linear ubiquitination induces NEMO phase separation to activate NF-κB signaling.
Life science alliance (2023-02-01)
Simran Goel, Rosario Oliva, Sadasivam Jeganathan, Verian Bader, Laura J Krause, Simon Kriegler, Isabelle D Stender, Chadwick W Christine, Ken Nakamura, Jan-Erik Hoffmann, Roland Winter, Jörg Tatzelt, Konstanze F Winklhofer
ABSTRACT
The NF-κB essential modulator NEMO is the core regulatory component of the inhibitor of κB kinase complex, which is a critical checkpoint in canonical NF-κB signaling downstream of innate and adaptive immune receptors. In response to various stimuli, such as TNF or IL-1β, NEMO binds to linear or M1-linked ubiquitin chains generated by LUBAC, promoting its oligomerization and subsequent activation of the associated kinases. Here we show that M1-ubiquitin chains induce phase separation of NEMO and the formation of NEMO assemblies in cells after exposure to IL-1β. Phase separation is promoted by both binding of NEMO to linear ubiquitin chains and covalent linkage of M1-ubiquitin to NEMO and is essential but not sufficient for its phase separation. Supporting the functional relevance of NEMO phase separation in signaling, a pathogenic NEMO mutant, which is impaired in both binding and linkage to linear ubiquitin chains, does not undergo phase separation and is defective in mediating IL-1β-induced NF-κB activation.
Sigma-Aldrich
Anti-IKBKG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Pricing and availability is not currently available.