Merck
  • Targeting cardiomyocyte ADAM10 ectodomain shedding promotes survival early after myocardial infarction.
Targeting cardiomyocyte ADAM10 ectodomain shedding promotes survival early after myocardial infarction.
Nature communications (2022-12-11)
Erik Klapproth, Anke Witt, Pauline Klose, Johanna Wiedemann, Nikitha Vavilthota, Stephan R Künzel, Susanne Kämmerer, Mario Günscht, David Sprott, Mathias Lesche, Fabian Rost, Andreas Dahl, Erik Rauch, Lars Kattner, Silvio Weber, Peter Mirtschink, Irakli Kopaliani, Kaomei Guan, Kristina Lorenz, Paul Saftig, Michael Wagner, Ali El-Armouche
ABSTRACT
After myocardial infarction the innate immune response is pivotal in clearing of tissue debris as well as scar formation, but exaggerated cytokine and chemokine secretion with subsequent leukocyte infiltration also leads to further tissue damage. Here, we address the value of targeting a previously unknown a disintegrin and metalloprotease 10 (ADAM10)/CX3CL1 axis in the regulation of neutrophil recruitment early after MI. We show that myocardial ADAM10 is distinctly upregulated in myocardial biopsies from patients with ischemia-driven cardiomyopathy. Intriguingly, upon MI in mice, pharmacological ADAM10 inhibition as well as genetic cardiomycyte-specific ADAM10 deletion improves survival with markedly enhanced heart function and reduced scar size. Mechanistically, abolished ADAM10-mediated CX3CL1 ectodomain shedding leads to diminished IL-1β-dependent inflammation, reduced neutrophil bone marrow egress as well as myocardial tissue infiltration. Thus, our data shows a conceptual insight into how acute MI induces chemotactic signaling via ectodomain shedding in cardiomyocytes.
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross-adsorbed (min X Rat), CF594 antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-ADAM 10 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
HL-1 Cardiac Muscle Cell Line, HL-1 Cardiac Muscle Cell Line has been extensively characterized and is a valuable model system to address questions of cardiac biology at the cellular & molecular levels.