Merck
  • The germline factor DDX4 contributes to the chemoresistance of small cell lung cancer cells.
The germline factor DDX4 contributes to the chemoresistance of small cell lung cancer cells.
Communications biology (2023-01-19)
Christopher Noyes, Shunsuke Kitajima, Fengkai Li, Yusuke Suita, Saradha Miriyala, Shakson Isaac, Nagib Ahsan, Erik Knelson, Amir Vajdi, Tetsuo Tani, Tran C Thai, Derek Xu, Junko Murai, Nikos Tapinos, Chiaki Takahashi, David A Barbie, Mamiko Yajima
ABSTRACT
Human cancers often re-express germline factors, yet their mechanistic role in oncogenesis and cancer progression remains unknown. Here we demonstrate that DEAD-box helicase 4 (DDX4), a germline factor and RNA helicase conserved in all multicellular organisms, contributes to increased cell motility and cisplatin-mediated drug resistance in small cell lung cancer (SCLC) cells. Proteomic analysis suggests that DDX4 expression upregulates proteins related to DNA repair and immune/inflammatory response. Consistent with these trends in cell lines, DDX4 depletion compromised in vivo tumor development while its overexpression enhanced tumor growth even after cisplatin treatment in nude mice. Further, the relatively higher DDX4 expression in SCLC patients correlates with decreased survival and shows increased expression of immune/inflammatory response markers. Taken together, we propose that DDX4 increases SCLC cell survival, by increasing the DNA damage and immune response pathways, especially under challenging conditions such as cisplatin treatment.
Sigma-Aldrich
Etoposide, A cell-permeable derivative of podophyllotoxin that acts as a topoisomerase II inhibitor (IC₅₀ = 59.2 µM) has major activity against a number of tumors, including germ cell neoplasms, small cell lung cancer, and malignant lymphoma.
Sigma-Aldrich
Hydroxyurea, Anti-neoplastic agent that blocks DNA replication by inhibiting ribonucleotide reductase.
Sigma-Aldrich
Cisplatin, Cisplatin - CAS 15663-27-1, is a platinum coordination complex with potent anti-neoplastic activity. Induces apoptosis in cancer cells, possibly via caspase-3 activation.
Sigma-Aldrich
Camptothecin, Camptotheca acuminata, A cell-permeable DNA topoisomerase I inhibitor.