- Induced Pluripotent Stem Cells: Generation, Characterization, and Differentiation--Methods and Protocols.
Induced Pluripotent Stem Cells: Generation, Characterization, and Differentiation--Methods and Protocols.
Methods in molecular biology (Clifton, N.J.) (2014-11-19)
Veronica Kon Graversen, Sai H Chavala
PMID25403469
ABSTRACT
Reprogramming fibroblasts into induced pluripotent stem cells (iPSC) remains a promising technique for cell replacement therapy. Diverse populations of somatic cells have been examined for their reprogramming potential. Recently, ocular ciliary body epithelial cells (CECs) have been reprogrammed with high reprogramming efficiency and single transcription factor reprogramming, making them an exciting candidate for cellular reprogramming strategies.
MATERIALS