Merck
  • Degradation of Ofloxacin by Perylene Diimide Supramolecular Nanofiber Sunlight-Driven Photocatalysis.
Degradation of Ofloxacin by Perylene Diimide Supramolecular Nanofiber Sunlight-Driven Photocatalysis.
Environmental science & technology (2019-01-04)
Ping Chen, Lee Blaney, Giovanni Cagnetta, Jun Huang, Bin Wang, Yujue Wang, Shubo Deng, Gang Yu
ABSTRACT
This study describes a promising sunlight-driven photocatalyst for the treatment of ofloxacin and other fluoroquinolone antibiotics in water and wastewater. Perylene diimide (PDI) supramolecular nanofibers, which absorb a broad spectrum of sunlight, were prepared via a facile acidification polymerization protocol. Under natural sunlight, the PDI photocatalysts achieved rapid treatment of fluoroquinolone antibiotics, including ciprofloxacin, enrofloxacin, norfloxacin, and ofloxacin. The fastest degradation was observed for ofloxacin, which had a half-life of 2.08 min for the investigated conditions. Various light sources emitting in the UV-vis spectrum were tested, and blue light was found to exhibit the fastest ofloxacin transformation kinetics due to the strong absorption by the PDI catalyst. Reactive species, namely, h+, 1O2, and O2•-, comprised the primary photocatalytic mechanisms for ofloxacin degradation. Frontier electron density calculations and mass spectrometry were used to verify the major degradation pathways of ofloxacin by the PDI-sunlight photocatalytic system and identify the transformation products of ofloxacin, respectively. Degradation mainly occurred through demethylation at the piperazine ring, ketone formation at the morpholine moiety, and aldehyde reaction at the piperazinyl group. An overall mechanism was proposed for ofloxacin degradation in the PDI-sunlight photocatalytic system, and the effects of water quality constituents were examined to determine performance in real water/wastewater systems. Ultimately, the aggregate results from this study highlight the suitability of the PDI-sunlight photocatalytic system to treat antibiotics in real water and wastewater systems.
Sigma-Aldrich
PTCDI