Merck
  • Minocycline mitigates the gliogenic effects of proinflammatory cytokines on neural stem cells.
Minocycline mitigates the gliogenic effects of proinflammatory cytokines on neural stem cells.
Journal of neuroscience research (2015-11-04)
Sabine Ulrike Vay, Stefan Blaschke, Rebecca Klein, Gereon Rudolf Fink, Michael Schroeter, Maria Adele Rueger
ABSTRACT
Mobilizing endogenous neural stem cells (NSCs) in the adult brain is designed to enhance the brain's regenerative capacity after cerebral lesions, e.g., as a result of stroke. Cerebral ischemia elicits neuroinflammatory processes affecting NSCs in multiple ways, the precise mechanisms of which currently remain elusive. An inhibitory effect of minocycline on microglia activation, a hallmark of postischemic neuroinflammation, has already been demonstrated in clinical trials, showing minocycline to be safe and potentially effective in ischemic stroke. Here we investigate the direct effects of minocycline and of proinflammatory cytokines on the differentiation potential of NSCs in vitro and in vivo. Primary fetal rat NSCs were treated with minocycline plus a combination of the proinflammatory cytokines tumor necrosis factor-α, interleukin 1β, and interleukin 6. The differentiation fate of NSCs was assessed immunocytochemically. To investigate the effects of minocycline and inflammation in vivo, minocycline or lipopolysaccharides were injected intraperitoneally into adult rats, with subsequent immunohistochemistry. Minocycline alone did not affect the differentiation potential of NSCs in vivo or in vitro. In contrast, proinflammatory cytokines accelerated the differentiation of NSCs, promoting an astrocytic fate while inhibiting neurogenesis in vitro and in vivo. It is interesting to note that minocycline counteracted this cytokine-induced rapid astrocytic differentiation and restored the neurogenic and oligodendrogliogenic potential of NSCs. Data suggest that minocycline antagonizes the rapid glial differentiation induced by proinflammatory cytokines following cerebral ischemia but without having a direct effect on the differentiation potential of NSCs. Thus, minocycline constitutes a promising drug for stroke research, counteracting the detrimental effects of postischemic neuroinflammation in multiple ways.
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
Anti-Doublecortin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®