Merck
Immunoprecipitation of Death Inducing Signaling Complex by Caspase-8.
Methods in molecular biology (Clifton, N.J.) (2017-01-13)
Andrei Alexandru Constantinescu, Aymeric Morlé, Olivier Micheau
ABSTRACT
Analysis of CD95/Fas complexes by immunoprecipitation has long relied on the monoclonal antibody APO1 or tagged recombinant Fas ligand. Immunoprecipitation is an elegant and efficient procedure to investigate endogenous protein interactions or complexes. Provided that the targeted complex is soluble in mild detergent these complexes can be recovered using protein A/G-coupled Sepharose beads and further analyzed after denaturation and electrophoretic separation by western blotting or mass spectrometry. Herein, we describe in detail the method used in our laboratory to immunoprecipitate and analyze by immunoblot complexes containing caspase-8, using a commercial antibody directed against caspase-8.
Sigma-Aldrich
Anti-DR5 Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Anti-DR4 Antibody, CT, Chemicon®, from rabbit