Merck
  • Design strategy for 25% external quantum efficiency in green and blue thermally activated delayed fluorescent devices.
Design strategy for 25% external quantum efficiency in green and blue thermally activated delayed fluorescent devices.
Advanced materials (Deerfield Beach, Fla.) (2015-08-27)
Dong Ryun Lee, Mounggon Kim, Sang Kyu Jeon, Seok-Ho Hwang, Chil Won Lee, Jun Yeob Lee
ABSTRACT
Carbazole- and triazine-derived thermally activated delayed fluorescent (TADF) emitters, with three donor units and an even distribution of the highest occupied molecular orbital, achieve high external quantum efficiencies of above 25% in blue and green TADF devices.
Sigma-Aldrich
TCzTrz, ≥99% (HPLC)