Merck
  • Controlled X-chromosome dynamics defines meiotic potential of female mouse in vitro germ cells.
Controlled X-chromosome dynamics defines meiotic potential of female mouse in vitro germ cells.
The EMBO journal (2022-05-24)
Jacqueline Severino, Moritz Bauer, Tom Mattimoe, Niccolò Arecco, Luca Cozzuto, Patricia Lorden, Norio Hamada, Yoshiaki Nosaka, So I Nagaoka, Pauline Audergon, Antonio Tarruell, Holger Heyn, Katsuhiko Hayashi, Mitinori Saitou, Bernhard Payer
ABSTRACT
The mammalian germline is characterized by extensive epigenetic reprogramming during its development into functional eggs and sperm. Specifically, the epigenome requires resetting before parental marks can be established and transmitted to the next generation. In the female germline, X-chromosome inactivation and reactivation are among the most prominent epigenetic reprogramming events, yet very little is known about their kinetics and biological function. Here, we investigate X-inactivation and reactivation dynamics using a tailor-made in vitro system of primordial germ cell-like cell (PGCLC) differentiation from mouse embryonic stem cells. We find that X-inactivation in PGCLCs in vitro and in germ cell-competent epiblast cells in vivo is moderate compared to somatic cells, and frequently characterized by escaping genes. X-inactivation is followed by step-wise X-reactivation, which is mostly completed during meiotic prophase I. Furthermore, we find that PGCLCs which fail to undergo X-inactivation or reactivate too rapidly display impaired meiotic potential. Thus, our data reveal fine-tuned X-chromosome remodelling as a critical feature of female germ cell development towards meiosis and oogenesis.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
RAD51-Stimulatory Compound-1, RS-1, A cell-permeable sulfonamido-benzamide-based allosteric regulator that stimulates DNA binding and recombinase activities of hRAD51 by locking hRAD51 in an active conformation without affecting its active site ATP hydrolysis.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra