Merck
  • Interplay mechanisms between progesterone and endocannabinoid receptors in regulating bull sperm capacitation and acrosome reaction.
Interplay mechanisms between progesterone and endocannabinoid receptors in regulating bull sperm capacitation and acrosome reaction.
Journal of cellular physiology (2022-04-28)
Abhishek Mahajan, Pratishtha Sharma, Abhishek K Mishra, Shashikant Gupta, Sarvajeet Yadav, Mukul Anand, Brijesh Yadav, Arun K Madan, Dilip K Swain
ABSTRACT
After ejaculation, sperm show a limited capacity for transcription and translation. In the oviduct, most of the signalling in sperm is nongenomic and is mediated through membrane receptors. Studies have shown that the cation channel of sperm (CatSper), cAMP, cGMP, protein kinases, and tyrosine phosphorylation are involved in the nongenomic signalling of progesterone (P4) in sperm. However, it is not known whether there is an interplay between P4 and cannabinoid receptors 1 and 2 (CB1 and CB2), transient receptor potential vanilloid 1 (TRPV1), CatSper channels, cAMP, inositol trisphosphate receptor (IP3R), and mitogen-activated protein kinase (MAPK); these potential regulators are involved in the regulation of capacitation and the acrosome reaction. In the present study, selective blockers of CB1, CB2, TRPV1, CatSper channels, cAMP, protein kinase A (PKA), IP3R, and MAPK were used to identify their involvement in P4-mediated bull sperm capacitation and the acrosome reaction. Selective blocking of any one of the molecules caused a significant reduction in P4 signalling (p < 0.05). Interestingly, blocking these molecules in combination followed by treatment with P4 resulted in the complete absence of capacitation and the acrosome reaction. Blocking a single receptor was not able to eliminate the P4-induced capacitation and the acrosome reaction. In addition to the CB1 and CB2 receptors, there may be other signalling pathways that mediate P4 signalling. In conclusion, P4 signalling exhibited interplay with the cannabinoid receptors. The regulation of sperm capacitation and the acrosome reaction also involved cAMP, PKA, l-type and T-type calcium channels, TRPV1, inositol trisphosphate, and MAPK.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Phosphotyrosine antibody, Mouse monoclonal, clone pT-154, purified from hybridoma cell culture