Merck
  • Cystic fibrosis transmembrane conductance regulator as a model substrate to study endoplasmic reticulum protein quality control in mammalian cells.
Cystic fibrosis transmembrane conductance regulator as a model substrate to study endoplasmic reticulum protein quality control in mammalian cells.
Methods in molecular biology (Clifton, N.J.) (2005-05-27)
J Michael Younger, Chun-Yang Fan, Liling Chen, Meredith F N Rosser, Cam Patterson, Douglas M Cyr
ABSTRACT
Components of the ubiquitin-proteasome system function on the surface of the endoplasmic reticulum (ER) to select misfolded proteins for degradation. Herein we describe methods that allow for the study of the pathway for proteasomal degradation of the cystic fibrosis transmembrane conductance regulator (CFTR). The experimental system described employs transiently transfected HEK-293 cells and is utilized to monitor the biogenesis of CFTR by Western blot and pulse-chase analysis.
Sigma-Aldrich
ALLN, Cell-permeable inhibitor of calpain I (Ki = 190 nM), calpain II (Ki = 220 nM), cathepsin B (Ki = 150 nM), and cathepsin L (Ki = 500 pM).
Sigma-Aldrich
Anti-CFTR Antibody, clone MM13-4, clone MM13-4, Upstate®, from mouse