Merck
  • Establishment of human fetal hepatocyte organoids and CRISPR-Cas9-based gene knockin and knockout in organoid cultures from human liver.
Establishment of human fetal hepatocyte organoids and CRISPR-Cas9-based gene knockin and knockout in organoid cultures from human liver.
Nature protocols (2020-11-29)
Delilah Hendriks, Benedetta Artegiani, Huili Hu, Susana Chuva de Sousa Lopes, Hans Clevers
ABSTRACT
The liver is composed of two epithelial cell types: hepatocytes and liver ductal cells. Culture conditions for expansion of human liver ductal cells in vitro as organoids were previously described in a protocol; however, primary human hepatocytes remained hard to expand, until recently. In this protocol, we provide full details of how we overcame this limitation, establishing culture conditions that facilitate long-term expansion of human fetal hepatocytes as organoids. In addition, we describe how to generate (multi) gene knockouts using CRISPR-Cas9 in both human fetal hepatocyte and adult liver ductal organoid systems. Using a CRISPR-Cas9 and homology-independent organoid transgenesis (CRISPR-HOT) approach, efficient gene knockin can be achieved in these systems. These gene knockin and knockout approaches, and their multiplexing, should be useful for a variety of applications, such as disease modeling, investigating gene functions and studying processes, such as cellular differentiation and cell division. The protocol to establish human fetal hepatocyte organoid cultures takes ~1-2 months. The protocols to genome engineer human liver ductal organoids and human fetal hepatocyte organoids take 2-3 months.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
[Leu15]-Gastrin I human, ≥95% (HPLC)
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone RCK105, clone RCK105, Chemicon®, from mouse
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture