Merck
  • LUBAC regulates ciliogenesis by promoting CP110 removal from the mother centriole.
LUBAC regulates ciliogenesis by promoting CP110 removal from the mother centriole.
The Journal of cell biology (2021-11-24)
Xiao-Lin Shen, Jin-Feng Yuan, Xuan-He Qin, Guang-Ping Song, Huai-Bin Hu, Hai-Qing Tu, Zeng-Qing Song, Pei-Yao Li, Yu-Ling Xu, Sen Li, Xiao-Xiao Jian, Jia-Ning Li, Chun-Yu He, Xi-Ping Yu, Li-Yun Liang, Min Wu, Qiu-Ying Han, Kai Wang, Ai-Ling Li, Tao Zhou, Yu-Cheng Zhang, Na Wang, Hui-Yan Li
ABSTRACT
Primary cilia transduce diverse signals in embryonic development and adult tissues. Defective ciliogenesis results in a series of human disorders collectively known as ciliopathies. The CP110-CEP97 complex removal from the mother centriole is an early critical step for ciliogenesis, but the underlying mechanism for this step remains largely obscure. Here, we reveal that the linear ubiquitin chain assembly complex (LUBAC) plays an essential role in ciliogenesis by targeting the CP110-CEP97 complex. LUBAC specifically generates linear ubiquitin chains on CP110, which is required for CP110 removal from the mother centriole in ciliogenesis. We further identify that a pre-mRNA splicing factor, PRPF8, at the distal end of the mother centriole acts as the receptor of the linear ubiquitin chains to facilitate CP110 removal at the initial stage of ciliogenesis. Thus, our study reveals a direct mechanism of regulating CP110 removal in ciliogenesis and implicates the E3 ligase LUBAC as a potential therapy target of cilia-associated diseases, including ciliopathies and cancers.
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Sharpin Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-TTBK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FBF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HOIL-1 Antibody, clone 2E2, clone 2E2, from mouse
Sigma-Aldrich
Anti-RPGRIP1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks