Merck
  • Transcription factors AP-2α and AP-2β regulate distinct segments of the distal nephron in the mammalian kidney.
Transcription factors AP-2α and AP-2β regulate distinct segments of the distal nephron in the mammalian kidney.
Nature communications (2022-04-27)
Joseph O Lamontagne, Hui Zhang, Alia M Zeid, Karin Strittmatter, Alicia D Rocha, Trevor Williams, Sheryl Zhang, Alexander G Marneros
ABSTRACT
Transcription factors AP-2α and AP-2β have been suggested to regulate the differentiation of nephron precursor populations towards distal nephron segments. Here, we show that in the adult mammalian kidney AP-2α is found in medullary collecting ducts, whereas AP-2β is found in distal nephron segments except for medullary collecting ducts. Inactivation of AP-2α in nephron progenitor cells does not affect mammalian nephrogenesis, whereas its inactivation in collecting ducts leads to defects in medullary collecting ducts in the adult. Heterozygosity for AP-2β in nephron progenitor cells leads to progressive distal convoluted tubule abnormalities and β-catenin/mTOR hyperactivation that is associated with renal fibrosis and cysts. Complete loss of AP-2β in nephron progenitor cells caused an absence of distal convoluted tubules, renal cysts, and fibrosis with β-catenin/mTOR hyperactivation, and early postnatal death. Thus, AP-2α and AP-2β have non-redundant distinct spatiotemporal functions in separate segments of the distal nephron in the mammalian kidney.
Sigma-Aldrich
Monoclonal Anti-Calbindin-D-28K antibody produced in mouse, clone CB-955, ascites fluid
Sigma-Aldrich
Anti-Water Channel Aquaporin 3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-TFAP2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Anti-Thiazide-Sensitive NaCl Cotransporter Antibody, Chemicon®, from rabbit