Merck
  • Evaluation of porcine urine-derived cells as nuclei donor for somatic cell nuclear transfer.
Evaluation of porcine urine-derived cells as nuclei donor for somatic cell nuclear transfer.
Journal of veterinary science (2022-04-02)
Yu-Ting Zhang, Wang Yao, Meng-Jia Chai, Wen-Jing Liu, Yan Liu, Zhong-Hua Liu, Xiao-Gang Weng
ABSTRACT
Somatic cell nuclear transfer (SCNT) is used widely in cloning, stem cell research, and regenerative medicine. The type of donor cells is a key factor affecting the SCNT efficiency. This study examined whether urine-derived somatic cells could be used as donors for SCNT in pigs. The viability of cells isolated from urine was assessed using trypan blue and propidium iodide staining. The H3K9me3/H3K27me3 level of the cells was analyzed by immunofluorescence. The in vitro developmental ability of SCNT embryos was evaluated by the blastocyst rate and the expression levels of the core pluripotency factor. Blastocyst cell apoptosis was examined using a terminal deoxynucleotidyl transferase dUTP nick end-labeling assay. The in vivo developmental ability of SCNT embryos was evaluated after embryo transfer. Most sow urine-derived cells were viable and could be cultured and propagated easily. On the other hand, most of the somatic cells isolated from the boar urine exhibited poor cellular activity. The in vitro development efficiency between the embryos produced by SCNT using porcine embryonic fibroblasts (PEFs) and urine-derived cells were similar. Moreover, The H3K9me3 in SCNT embryos produced from sow urine-derived cells and PEFs at the four-cell stage showed similar intensity. The levels of Oct4, Nanog, and Sox2 expression in blastocysts were similar in the two groups. Furthermore, there is a similar apoptotic level of cloned embryos produced by the two types of cells. Finally, the full-term development ability of the cloned embryos was evaluated, and the cloned fetuses from the urine-derived cells showed absorption. Sow urine-derived cells could be used to produce SCNT embryos.
Sigma-Aldrich
Anti-NANOG antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit