Merck
  • Inhibiting pathologically active ADAM10 rescues synaptic and cognitive decline in Huntington's disease.
Inhibiting pathologically active ADAM10 rescues synaptic and cognitive decline in Huntington's disease.
The Journal of clinical investigation (2019-05-08)
Elena Vezzoli, Ilaria Caron, Francesca Talpo, Dario Besusso, Paola Conforti, Elisa Battaglia, Elisa Sogne, Andrea Falqui, Lara Petricca, Margherita Verani, Paola Martufi, Andrea Caricasole, Alberto Bresciani, Ottavia Cecchetti, Pia Rivetti di Val Cervo, Giulio Sancini, Olaf Riess, Hoa Nguyen, Lisa Seipold, Paul Saftig, Gerardo Biella, Elena Cattaneo, Chiara Zuccato
ABSTRACT
A disintegrine and metalloproteinase 10 (ADAM10) is implicated in synaptic function through its interaction with postsynaptic receptors and adhesion molecules. Here, we report that levels of active ADAM10 are increased in Huntington's disease (HD) mouse cortices and striata and in human postmortem caudate. We show that, in the presence of polyglutamine-expanded (polyQ-expanded) huntingtin (HTT), ADAM10 accumulates at the postsynaptic densities (PSDs) and causes excessive cleavage of the synaptic protein N-cadherin (N-CAD). This aberrant phenotype is also detected in neurons from HD patients where it can be reverted by selective silencing of mutant HTT. Consistently, ex vivo delivery of an ADAM10 synthetic inhibitor reduces N-CAD proteolysis and corrects electrophysiological alterations in striatal medium-sized spiny neurons (MSNs) of 2 HD mouse models. Moreover, we show that heterozygous conditional deletion of ADAM10 or delivery of a competitive TAT-Pro-ADAM10709-729 peptide in R6/2 mice prevents N-CAD proteolysis and ameliorates cognitive deficits in the mice. Reduction in synapse loss was also found in R6/2 mice conditionally deleted for ADAM10. Taken together, these results point to a detrimental role of hyperactive ADAM10 at the HD synapse and provide preclinical evidence of the therapeutic potential of ADAM10 inhibition in HD.
Sigma-Aldrich
1(S),9(R)-(−)-Bicuculline methiodide, ≥95.0% (HPCE)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
ESGRO Complete Accutase, The ESGRO Complete Accutase is a cell detachment solution of proteolytic & collagenolytic enzymes, qualified for use for the detachment of mouse embryonic stem cells cultured in serum-free conditions with ESGRO Complete Clonal Grade Medium.