Merck
  • Targeting necroptosis in muscle fibers ameliorates inflammatory myopathies.
Targeting necroptosis in muscle fibers ameliorates inflammatory myopathies.
Nature communications (2022-01-12)
Mari Kamiya, Fumitaka Mizoguchi, Kimito Kawahata, Dengli Wang, Masahiro Nishibori, Jessica Day, Cynthia Louis, Ian P Wicks, Hitoshi Kohsaka, Shinsuke Yasuda
ABSTRACT
Muscle cell death in polymyositis is induced by CD8+ cytotoxic T lymphocytes. We hypothesized that the injured muscle fibers release pro-inflammatory molecules, which would further accelerate CD8+ cytotoxic T lymphocytes-induced muscle injury, and inhibition of the cell death of muscle fibers could be a novel therapeutic strategy to suppress both muscle injury and inflammation in polymyositis. Here, we show that the pattern of cell death of muscle fibers in polymyositis is FAS ligand-dependent necroptosis, while that of satellite cells and myoblasts is perforin 1/granzyme B-dependent apoptosis, using human muscle biopsy specimens of polymyositis patients and models of polymyositis in vitro and in vivo. Inhibition of necroptosis suppresses not only CD8+ cytotoxic T lymphocytes-induced cell death of myotubes but also the release of inflammatory molecules including HMGB1. Treatment with a necroptosis inhibitor or anti-HMGB1 antibodies ameliorates myositis-induced muscle weakness as well as muscle cell death and inflammation in the muscles. Thus, targeting necroptosis in muscle cells is a promising strategy for treating polymyositis providing an alternative to current therapies directed at leukocytes.
Sigma-Aldrich
Mouse IgM Negative Control, clone GC323, Mouse IgM Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.
Sigma-Aldrich
Anti-Caspase-8 (active form p18 subunit) Antibody, clone 2B12.1, clone 2B12.1, from mouse