Merck
  • The human brain somatostatin interactome: SST binds selectively to P-type family ATPases.
The human brain somatostatin interactome: SST binds selectively to P-type family ATPases.
PloS one (2019-05-29)
Michael Solarski, Declan Williams, Mohadeseh Mehrabian, Hansen Wang, Holger Wille, Gerold Schmitt-Ulms
ABSTRACT
Somatostatin (SST) is a cyclic peptide that is understood to inhibit the release of hormones and neurotransmitters from a variety of cells by binding to one of five canonical G protein-coupled SST receptors (SSTR1 to SSTR5). Recently, SST was also observed to interact with the amyloid beta (Aβ) peptide and affect its aggregation kinetics, raising the possibility that it may bind other brain proteins. Here we report on an SST interactome analysis that made use of human brain extracts as biological source material and incorporated advanced mass spectrometry workflows for the relative quantitation of SST binding proteins. The analysis revealed SST to predominantly bind several members of the P-type family of ATPases. Subsequent validation experiments confirmed an interaction between SST and the sodium-potassium pump (Na+/K+-ATPase) and identified a tryptophan residue within SST as critical for binding. Functional analyses in three different cell lines indicated that SST might negatively modulate the K+ uptake rate of the Na+/K+-ATPase.
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
ReNcell VM Human Neural Progenitor Cell LIne, ReNcell VM is an immortalized human neural progenitor cell line with the ability to readily differentiate into neurons & glial cells.
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial