Merck
  • Mapping Cell Membrane Organization and Dynamics Using Soft Nanoimprint Lithography.
Mapping Cell Membrane Organization and Dynamics Using Soft Nanoimprint Lithography.
ACS applied materials & interfaces (2020-05-29)
T Sansen, D Sanchez-Fuentes, R Rathar, A Colom-Diego, F El Alaoui, J Viaud, M Macchione, S de Rossi, S Matile, R Gaudin, V Bäcker, A Carretero-Genevrier, L Picas
ABSTRACT
Membrane shape is a key feature of many cellular processes, including cell differentiation, division, migration, and trafficking. The development of nanostructured surfaces allowing for the in situ manipulation of membranes in living cells is crucial to understand these processes, but this requires complicated and limited-access technologies. Here, we investigate the self-organization of cellular membranes by using a customizable and benchtop method allowing one to engineer 1D SiO2 nanopillar arrays of defined sizes and shapes on high-performance glass compatible with advanced microscopies. As a result of this original combination, we provide a mapping of the morphology-induced modulation of the cell membrane mechanics, dynamics and steady-state organization of key protein complexes implicated in cellular trafficking and signal transduction.
Sigma-Aldrich
Cytochalasin D, from Zygosporium mansonii, ≥98% (TLC and HPLC), powder
Sigma-Aldrich
Jasplakinolide, ≥97% (HPLC)
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-CLTA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone MB1.2, clone MB1.2, Chemicon®, from rat