Merck
  • Evaluation of Amikacin Pharmacokinetics and Pharmacodynamics for Optimal Initial Dosing Regimen.
Evaluation of Amikacin Pharmacokinetics and Pharmacodynamics for Optimal Initial Dosing Regimen.
Drugs in R&D (2017-01-08)
Hideo Kato, Mao Hagihara, Jun Hirai, Daisuke Sakanashi, Hiroyuki Suematsu, Naoya Nishiyama, Yusuke Koizumi, Yuka Yamagishi, Katsuhiko Matsuura, Hiroshige Mikamo
ABSTRACT
Amikacin has been one of the important antimicrobial agents against Gram-negative pathogens. However, there is discrepancy regarding the amikacin initial dosage, with some reports recently recommending ≥25 mg/kg and others the conventional dosage (15-20 mg/kg). Hence, we evaluated the optimal initial dosing regimen of amikacin. Pharmacokinetic (PK) parameters were estimated using a population PK analysis. The pharmacodynamic (PD) target was a ratio of ≥8 between the concentration achieved 1 h after beginning the infusion (C peak) and the minimal inhibitory concentration (MIC) of the liable bacteria. Based on the population PK parameters, we simulated individual C peak for several dosing regimens by Monte Carlo method and analyzed the C peak/MIC ratio for MICs from 0.5 to 32 μg/mL. This study included 35 infected patients (25 males), with a median (range) age and body weight of 70 (15-95) years and 49.5 (32.5-78) kg, respectively. A two-compartment model was used, and total body clearance (CL) significantly correlated with creatinine clearance, and volume of distribution (V d) with body weight. Regarding the probability to achieve a C peak/MIC of ≥8, the 15 mg/kg regimen was sufficient to achieve the PK/PD target in ≥90% of patients for a MIC of 4 μg/mL or less. The cumulative fraction of response in Pseudomonas aeruginosa was that 76% of patients achieved a C peak/MIC of 8 with the amikacin dosage of 15 mg/kg/day. We suggest that the 15-mg/kg once-daily dosage of amikacin be recommended as the initial dosage. As its maintenance dosage, the 15 mg/kg/day amikacin dosage is needed for a MIC of ≤4 μg/mL, and amikacin monotherapy for a MIC of ≥8 μg/mL should be avoided.
Sigma-Aldrich
Amikacin Ready Made Solution, 25 mg/mL in water
Pricing and availability is not currently available.