Merck
  • Intrinsic bias at non-canonical, β-arrestin-coupled seven transmembrane receptors.
Intrinsic bias at non-canonical, β-arrestin-coupled seven transmembrane receptors.
Molecular cell (2021-09-29)
Shubhi Pandey, Punita Kumari, Mithu Baidya, Ryoji Kise, Yubo Cao, Hemlata Dwivedi-Agnihotri, Ramanuj Banerjee, Xaria X Li, Cedric S Cui, John D Lee, Kouki Kawakami, Jagannath Maharana, Ashutosh Ranjan, Madhu Chaturvedi, Gagan Deep Jhingan, Stéphane A Laporte, Trent M Woodruff, Asuka Inoue, Arun K Shukla
ABSTRACT
G-protein-coupled receptors (GPCRs), also known as seven transmembrane receptors (7TMRs), typically interact with two distinct signal-transducers, i.e., G proteins and β-arrestins (βarrs). Interestingly, there are some non-canonical 7TMRs that lack G protein coupling but interact with βarrs, although an understanding of their transducer coupling preference, downstream signaling, and structural mechanism remains elusive. Here, we characterize two such non-canonical 7TMRs, namely, the decoy D6 receptor (D6R) and the complement C5a receptor subtype 2 (C5aR2), in parallel with their canonical GPCR counterparts. We discover that D6R and C5aR2 efficiently couple to βarrs, exhibit distinct engagement of GPCR kinases (GRKs), and activate non-canonical downstream signaling pathways. We also observe that βarrs adopt distinct conformations for D6R and C5aR2, compared to their canonical GPCR counterparts, in response to common natural agonists. Our study establishes D6R and C5aR2 as βarr-coupled 7TMRs and provides key insights into their regulation and signaling with direct implication for biased agonism.
Sigma-Aldrich
DAPI, for nucleic acid staining
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
pIMAGO-biotin Phosphoprotein Detection Kit, for western blot, HRP-based detection (ECL)
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
U0126, U0126 is a elective inhibitor of MEK-1 and MEK-2; weak inhibitor of PKC, Raf, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, Abl, Cdk2 and Cdk4.