Merck
  • Short Communication: Quantification of the Effect of Mycotoxin Binders on the Bioavailability of Fat-Soluble Vitamins In Vitro.
Short Communication: Quantification of the Effect of Mycotoxin Binders on the Bioavailability of Fat-Soluble Vitamins In Vitro.
Animals : an open access journal from MDPI (2021-08-28)
Abdelhacib Kihal, María Ercilda Rodríguez-Prado, Carles Cristofol, Sergio Calsamiglia
ABSTRACT
The aim of this study was to determine the capacity of six mycotoxin binders (MTBs) to adsorb vitamins A, D and E in an in vitro system that simulates gastric and intestinal digestion. Experiment 1 evaluated the recovery rate of vitamins A, D and E in the incubation conditions. In Experiment 2, the main factors were the MTB (bentonite, clinoptilolite, sepiolite, montmorillonite, active carbon and yeast cell walls), vitamins (A, D and E) and incubation type (vitamins incubated separately or together). The recovery was high for vitamin D (83%) and E (93%), but low for vitamin A (23%), for which no further analyses were conducted. When incubated separately, vitamin D was only adsorbed by yeast cell wall (20.2%). Vitamin E adsorption was highest with bentonite (54.5%) and montmorillonite (46.3%) and lowest with sepiolite (16.6%) and active carbon (18.5%). When incubated together, vitamin D was not adsorbed by any MTB. Vitamin E adsorption was highest in bentonite (61.8%) and montmorillonite (50.7%) and lowest in sepiolite (15.4%). Results indicate that the bioavailability of vitamin E, but not that of vitamin D, may be reduced in the presence of MTBs.