Merck
  • Caucasian Gentiana Species: Untargeted LC-MS Metabolic Profiling, Antioxidant and Digestive Enzyme Inhibiting Activity of Six Plants.
Caucasian Gentiana Species: Untargeted LC-MS Metabolic Profiling, Antioxidant and Digestive Enzyme Inhibiting Activity of Six Plants.
Metabolites (2019-11-11)
Daniil N Olennikov, Aydan I Gadimli, Javanshir I Isaev, Nina I Kashchenko, Alexey S Prokopyev, Tatyana N Kataeva, Nadezhda K Chirikova, Cecile Vennos
ABSTRACT
The members of Gentiana genus are widely distributed in the Caucasus region where they are used as phytoremedies, but they still have not been studied for their chemical composition and bioactivity. High-performance liquid chromatography with diode array and electrospray triple quadrupole mass detection (HPLC-DAD-ESI-QQQ-MS) was used to investigate metabolites of herb and roots of six gentians (Gentiana asclepiadea, G. cruciata, G. gelida, G. paradoxa, G. pneumonanthe, G. septemfida) grown in the Caucasus. In total, 137 compounds were found including three carbohydrates, 71 iridoid glycosides (mostly loganic acid), loganin, swertiamarin, gentiopicroside and sweroside derivatives, 40 flavones C-, O-, C,O-glycosides (such as luteolin, apigenin, chrysoeriol, and acacetin derivatives), two phenolic O-glycosides, five hydroxycinnamates, eight xanthones, and seven triterpene glycosides. Most of these compounds were identified in gentian samples for the first time. Quantitative differences were found in levels of seven iridoid glycosides, nine glycosylflavones, and two xanthones obtained by HPLC-DAD assay. The gentian extracts were evaluated for their radical-scavenging properties against DPPH and superoxide anion radicals, lipid peroxidation inhibition, and α-amylase/α-glycosidase inhibition. The herb extracts showed higher activity than root extracts. Positive correlations were found between the content of quantified phenolics and antioxidant and digestive enzymes inhibiting activity. The findings presented in our work suggest that the Caucasian gentians are a good source of bioactive phytocompounds with antioxidant and antidiabetic potential.
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Gentiopicroside, ≥98% (HPLC)