Merck
  • Remodeling of dermal adipose tissue alleviates cutaneous toxicity induced by anti-EGFR therapy.
Remodeling of dermal adipose tissue alleviates cutaneous toxicity induced by anti-EGFR therapy.
eLife (2022-03-25)
Leying Chen, Qing You, Min Liu, Shuaihu Li, Zhaoyu Wu, Jiajun Hu, Yurui Ma, Liangyong Xia, Ying Zhou, Nan Xu, Shiyi Zhang
ABSTRACT
Anti-epidermal growth factor receptor (EGFR) therapy-associated cutaneous toxicity is a syndrome characterized by papulopustular rash, local inflammation, folliculitis, and microbial infection, resulting in a decrease in quality of life and dose interruption. However, no effective clinical intervention is available for this adverse effect. Here, we report the atrophy of dermal white adipose tissue (dWAT), a highly plastic adipose tissue with various skin-specific functions, correlates with rash occurrence and exacerbation in a murine model of EGFR inhibitor-induced rash. The reduction in dWAT is due to the inhibition of adipogenic differentiation by defects in peroxisome proliferator-activated receptor γ (PPARγ) signaling, and increased lipolysis by the induced expression of the lipolytic cytokine IL6. The activation of PPARγ by rosiglitazone maintains adipogenic differentiation and represses the transcription of IL6, eventually improving skin functions and ameliorating the severity of rash without altering the antitumor effects. Thus, activation of PPARγ represents a promising approach to ameliorate cutaneous toxicity in patients with cancer who receive anti-EGFR therapy.
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
Insulin, Zinc, Human, Recombinant, P. pastoris
Roche
Liberase TL Research Grade, lyophilized, suitable for tissue processing, optimum pH 7.4