Merck
  • High Fructose Drives the Serine Synthesis Pathway in Acute Myeloid Leukemic Cells.
High Fructose Drives the Serine Synthesis Pathway in Acute Myeloid Leukemic Cells.
Cell metabolism (2020-12-29)
Sangmoo Jeong, Angela Maria Savino, Rachel Chirayil, Ersilia Barin, Yuanming Cheng, Sun-Mi Park, Alexandra Schurer, Edouard Mullarky, Lewis C Cantley, Michael G Kharas, Kayvan R Keshari
ABSTRACT
A significant increase in dietary fructose consumption has been implicated as a potential driver of cancer. Metabolic adaptation of cancer cells to utilize fructose confers advantages for their malignant growth, but compelling therapeutic targets have not been identified. Here, we show that fructose metabolism of leukemic cells can be inhibited by targeting the de novo serine synthesis pathway (SSP). Leukemic cells, unlike their normal counterparts, become significantly dependent on the SSP in fructose-rich conditions as compared to glucose-rich conditions. This metabolic program is mediated by the ratio of redox cofactors, NAD+/NADH, and the increased SSP flux is beneficial for generating alpha-ketoglutarate from glutamine, which allows leukemic cells to proliferate even in the absence of glucose. Inhibition of PHGDH, a rate-limiting enzyme in the SSP, dramatically reduces leukemia engraftment in mice in the presence of high fructose, confirming the essential role of the SSP in the metabolic plasticity of leukemic cells.
Sigma-Aldrich
Anti-PHGDH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Ketohexokinase Inhibitor
Sigma-Aldrich
Pyruvate Kinase from rabbit muscle, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
L-Lactate Dehydrogenase (LDHA), from human, recombinant, expressed in E. coli, aqueous solution