Merck
  • Separate basolateral and apical phosphatidylcholine secretion routes in intestinally differentiated tumor cells.
Separate basolateral and apical phosphatidylcholine secretion routes in intestinally differentiated tumor cells.
World journal of gastroenterology (2009-12-10)
Daniel Gotthardt, Annika Braun, Anke Tietje, Karl Heinz Weiss, Robert Ehehalt, Wolfgang R Stremmel
ABSTRACT
To investigate whether the secretion of phosphatidylcholine (PC) in intestinal mucus occurs by apical secretion or via basolateral excretion and to determine its subsequent passage across the tight junctions to the apical mucus. We addressed this question using the polarized intestinally differentiated tumor cell line CaCo-2 grown on filters to confluence in Transwell culture chambers. The released PC and sphingomyelin (Sph) from apical and basolateral media were analyzed by mass spectrometry. The secreted PC species were identical in both compartments indicating the same intracellular origin of PC. However, PC secretion into the basolateral compartment was more effective, and the PC:Sph ratio in the basolateral compartment was significantly higher than that in the apical compartment (8.18 +/- 1.84 vs 4.31 +/- 1.22, P = 0.01). Both pathways were temperature sensitive and were unaltered in the presence of cyclosporine. The data demonstrate the PC secretion capacity of CaCo-2 cells and indicate two separated apical and basolateral release mechanisms.
Sigma-Aldrich
Bovine Serum Albumin, Cohn Fraction V, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially γ-globulin free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, protease, essentially free, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, Essentially fatty acid free
Sigma-Aldrich
Bovine Serum Albumin, dimer mol wt ~132 kDa
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially IgG-free, low endotoxin, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, low endotoxin, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, suitable for (for molecular biology), Non-acetylated
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, γ-irradiated, Globulin Free, BioXtra, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, suitable for RIA, pH 5.2, ≥96%
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, low endotoxin, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially fatty acid free and essentially globulin free, ≥99% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Bovine Serum Albumin, powder, BioXtra
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)