Merck
  • β-sitosterol reduces anxiety and synergizes with established anxiolytic drugs in mice.
β-sitosterol reduces anxiety and synergizes with established anxiolytic drugs in mice.
Cell reports. Medicine (2021-06-08)
Nicolas Panayotis, Philip A Freund, Letizia Marvaldi, Tali Shalit, Alexander Brandis, Tevie Mehlman, Michael M Tsoory, Mike Fainzilber
ABSTRACT
Anxiety and stress-related conditions represent a significant health burden in modern society. Unfortunately, most anxiolytic drugs are prone to side effects, limiting their long-term usage. Here, we employ a bioinformatics screen to identify drugs for repurposing as anxiolytics. Comparison of drug-induced gene-expression profiles with the hippocampal transcriptome of an importin α5 mutant mouse model with reduced anxiety identifies the hypocholesterolemic agent β-sitosterol as a promising candidate. β-sitosterol activity is validated by both intraperitoneal and oral application in mice, revealing it as the only clear anxiolytic from five closely related phytosterols. β-sitosterol injection reduces the effects of restraint stress, contextual fear memory, and c-Fos activation in the prefrontal cortex and dentate gyrus. Moreover, synergistic anxiolysis is observed when combining sub-efficacious doses of β-sitosterol with the SSRI fluoxetine. These preclinical findings support further development of β-sitosterol, either as a standalone anxiolytic or in combination with low-dose SSRIs.
Sigma-Aldrich
(−)-Epinephrine (+)-bitartrate salt, solid
Sigma-Aldrich
Fucosterol, ≥93%
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Fluoxetine hydrochloride, solid