Merck
Controlled functionalization of graphene oxide with sodium azide.
Nanoscale (2013-10-29)
Siegfried Eigler, Yichen Hu, Yoshitaka Ishii, Andreas Hirsch
ABSTRACT
We present the first example of azide functionalization on the surface of graphene oxide (GO), which preserves thermally unstable groups in GO through the mild reaction with sodium azide in solids. Experimental evidence, by (15)N solid-state NMR and other spectroscopic methods, indicates the substitution of organosulfate with azide anions as the reaction mechanism.
Sigma-Aldrich
Sodium azide-1-15N, (terminal N), 98 atom % 15N
Pricing and availability is not currently available.