Merck
  • An essential role of the autophagy activating kinase ULK1 in snRNP biogenesis.
An essential role of the autophagy activating kinase ULK1 in snRNP biogenesis.
Nucleic acids research (2021-06-08)
Katharina Schmitz, Jan Cox, Lea Marie Esser, Martin Voss, Katja Sander, Antje Löffler, Frank Hillebrand, Steffen Erkelenz, Heiner Schaal, Thilo Kähne, Stefan Klinker, Tao Zhang, Luitgard Nagel-Steger, Dieter Willbold, Sabine Seggewiß, David Schlütermann, Björn Stork, Matthias Grimmler, Sebastian Wesselborg, Christoph Peter
ABSTRACT
The biogenesis of small uridine-rich nuclear ribonucleoproteins (UsnRNPs) depends on the methylation of Sm proteins catalyzed by the methylosome and the subsequent action of the SMN complex, which assembles the heptameric Sm protein ring onto small nuclear RNAs (snRNAs). In this sophisticated process, the methylosome subunit pICln (chloride conductance regulatory protein) is attributed to an exceptional key position as an 'assembly chaperone' by building up a stable precursor Sm protein ring structure. Here, we show that-apart from its autophagic role-the Ser/Thr kinase ULK1 (Uncoordinated [unc-51] Like Kinase 1) functions as a novel key regulator in UsnRNP biogenesis by phosphorylation of the C-terminus of pICln. As a consequence, phosphorylated pICln is no longer capable to hold up the precursor Sm ring structure. Consequently, inhibition of ULK1 results in a reduction of efficient UsnRNP core assembly. Thus ULK1, depending on its complex formation, exerts different functions in autophagy or snRNP biosynthesis.
Sigma-Aldrich
Anti-SNRPF antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-ATG13 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
ULK1 (1-649), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
PRMT5/MEP50 Active human, recombinant, expressed in FreeStyle 293-F cells, ≥60% (SDS-PAGE)
Sigma-Aldrich
Anti-Atg101 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-SmB antibody, Mouse Monoclonal, clone 12F5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-SMN Antibody, clone 2B1, clone 2B1, from mouse
Sigma-Aldrich
Anti-SNRPG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution