Merck
  • Human alveolar type 2 epithelium transdifferentiates into metaplastic KRT5+ basal cells.
Human alveolar type 2 epithelium transdifferentiates into metaplastic KRT5+ basal cells.
Nature cell biology (2022-01-01)
Jaymin J Kathiriya, Chaoqun Wang, Minqi Zhou, Alexis Brumwell, Monica Cassandras, Claude Jourdan Le Saux, Max Cohen, Kostantinos-Dionysios Alysandratos, Bruce Wang, Paul Wolters, Michael Matthay, Darrell N Kotton, Harold A Chapman, Tien Peng
ABSTRACT
Loss of alveolar type 2 cells (AEC2s) and the ectopic appearance of basal cells in the alveoli characterize severe lung injuries such as idiopathic pulmonary fibrosis (IPF). Here we demonstrate that human alveolar type 2 cells (hAEC2s), unlike murine AEC2s, transdifferentiate into basal cells in response to fibrotic signalling in the lung mesenchyme, in vitro and in vivo. Single-cell analysis of normal hAEC2s and mesenchymal cells in organoid co-cultures revealed the emergence of pathologic fibroblasts and basaloid cells previously described in IPF. Transforming growth factor-β1 and anti-bone morphogenic protein signalling in the organoids promoted transdifferentiation. Trajectory and histologic analyses of both hAEC2-derived organoids and IPF epithelium indicated that hAEC2s transdifferentiate into basal cells through alveolar-basal intermediates that accumulate in proximity to pathologic CTHRC1hi/TGFB1hi fibroblasts. Our study indicates that hAEC2 loss and expansion of alveolar metaplastic basal cells in severe human lung injuries are causally connected through an hAEC2-basal cell lineage trajectory driven by aberrant mesenchyme.
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-phospho Smad1/Smad5/Smad8 (Ser463/465), from rabbit, purified by affinity chromatography
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone LL002, clone LL002, Chemicon®, from mouse
Sigma-Aldrich
Anti-Prosurfactant Protein C (proSP-C) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-MMP-7 Antibody, clone 141-7B2, clone 141-7B2, Chemicon®, from mouse