Merck
  • Extracellular Vesicles from Pancreatic Cancer Stem Cells Lead an Intratumor Communication Network (EVNet) to fuel tumour progression.
Extracellular Vesicles from Pancreatic Cancer Stem Cells Lead an Intratumor Communication Network (EVNet) to fuel tumour progression.
Gut (2022-01-12)
Carolina F Ruivo, Nuno Bastos, Barbara Adem, Ines Batista, Cecilia Duraes, Carlos A Melo, Stephanie A Castaldo, Francisco Campos-Laborie, Pedro Moutinho-Ribeiro, Barbara Morão, Ana Costa-Pinto, Soraia Silva, Hugo Osorio, Sergio Ciordia, Jose Luis Costa, David Goodrich, Bruno Cavadas, Luisa Pereira, Tony Kouzarides, Guilherme Macedo, Rui Maio, Fatima Carneiro, Marília Cravo, Raghu Kalluri, Jose Carlos Machado, Sonia A Melo
ABSTRACT
Intratumor heterogeneity drives cancer progression and therapy resistance. However, it has yet to be determined whether and how subpopulations of cancer cells interact and how this interaction affects the tumour. We have studied the spontaneous flow of extracellular vesicles (EVs) between subpopulations of cancer cells: cancer stem cells (CSC) and non-stem cancer cells (NSCC). To determine the biological significance of the most frequent communication route, we used pancreatic ductal adenocarcinoma (PDAC) orthotopic models, patient-derived xenografts (PDXs) and genetically engineered mouse models (GEMMs). We demonstrate that PDAC tumours establish an organised communication network between subpopulations of cancer cells using EVs called the EVNet). The EVNet is plastic and reshapes in response to its environment. Communication within the EVNet occurs preferentially from CSC to NSCC. Inhibition of this communication route by impairing Rab27a function in orthotopic xenographs, GEMMs and PDXs is sufficient to hamper tumour growth and phenocopies the inhibition of communication in the whole tumour. Mechanistically, we provide evidence that CSC EVs use agrin protein to promote Yes1 associated transcriptional regulator (YAP) activation via LDL receptor related protein 4 (LRP-4). Ex vivo treatment of PDXs with antiagrin significantly impairs proliferation and decreases the levels of activated YAP.Patients with high levels of agrin and low inactive YAP show worse disease-free survival. In addition, patients with a higher number of circulating agrin+ EVs show a significant increased risk of disease progression. PDAC tumours establish a cooperation network mediated by EVs that is led by CSC and agrin, which allows tumours to adapt and thrive. Targeting agrin could make targeted therapy possible for patients with PDAC and has a significant impact on CSC that feeds the tumour and is at the centre of therapy resistance.
Sigma-Aldrich
Monoclonal Anti-CD326-FITC antibody produced in mouse, clone VU-1D9, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-CD9 antibody produced in mouse, clone MEM-61, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)
Sigma-Aldrich
Anti-RAB27A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Nexinhib20, ≥98% (HPLC)
Sigma-Aldrich
Anti-Agrin Antibody, Chemicon®, from mouse