MilliporeSigma
  • Human iPSC-derived fallopian tube organoids with BRCA1 mutation recapitulate early-stage carcinogenesis.
Human iPSC-derived fallopian tube organoids with BRCA1 mutation recapitulate early-stage carcinogenesis.
Cell reports (2021-12-30)
Nur Yucer, Rodney Ahdoot, Michael J Workman, Alexander H Laperle, Maria S Recouvreux, Kathleen Kurowski, Diana J Naboulsi, Victoria Liang, Ying Qu, Jasmine T Plummer, Simon A Gayther, Sandra Orsulic, Beth Y Karlan, Clive N Svendsen
ABSTRACT
Germline pathogenic mutations in BReast CAncer (BRCA1) genes are thought to drive normal fallopian tube epithelial (FTE) cell transformation to high-grade serous ovarian cancer. No human models capture the sequence of events for disease initiation and progression. Here, we generate induced pluripotent stem cells (iPSCs) from healthy individuals and young ovarian cancer patients with germline pathogenic BRCA1 mutations (BRCA1mut). Following differentiation into FTE organoids, BRCA1mut lines exhibit cellular abnormalities consistent with neoplastic transformation compared to controls. BRCA1mut organoids show an increased production of cancer-specific proteins and survival following transplantation into mice. Organoids from women with the most aggressive ovarian cancer show the greatest pathology, indicating the potential value to predict clinical severity prior to disease onset. These human FTE organoids from BRCA1mut carriers provide a faithful physiological in vitro model of FTE lesion generation and early carcinogenesis. This platform can be used for personalized mechanistic and drug screening studies.
Sigma-Aldrich
Leukemia Inhibitory Factor human, 10 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-OVGP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-BRCA1 (Ab-1) Mouse mAb (MS110), liquid, clone MS110, Calbiochem®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, FITC conjugate, clone JBW301, Upstate®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MS-SAFE Protease and Phosphatase Inhibitor