Merck
  • Investigational therapeutics targeting the IL-4/IL-13/STAT-6 pathway for the treatment of asthma.
Investigational therapeutics targeting the IL-4/IL-13/STAT-6 pathway for the treatment of asthma.
European respiratory review : an official journal of the European Respiratory Society (2010-10-20)
C K Oh, G P Geba, N Molfino
ABSTRACT
Asthma is a complex, persistent, inflammatory disease characterised by airway hyperresponsiveness in association with airway inflammation. Studies suggest that regular use of high-dose inhaled corticosteroids and long-acting bronchodilators or omalizumab (a humanised monoclonal antibody that binds to immunoglobulin E and is often used as next-step therapy) may not be sufficient to provide asthma control in all patients, highlighting an important unmet need. Interleukin-4, interleukin-13, and the signal transducer and activator of transcription factor-6 are key components in the development of airway inflammation, mucus production, and airway hyperresponsiveness in asthma. Biological compounds targeting these molecules may provide a new therapeutic modality for patients with uncontrolled severe asthma. The purpose of this review is to summarise current studies of compounds targeting the interleukin-4/interleukin-13 pathway and to provide a rationale for the development of such compounds for this use.
Product Description
OX-81, Reactivity: Anti-Interleukin-4, rat, 95040518
Sigma-Aldrich
Anti-IL4 antibody produced in rabbit, affinity isolated antibody