Merck
  • STING regulates peripheral nerve regeneration and colony stimulating factor 1 receptor (CSF1R) processing in microglia.
STING regulates peripheral nerve regeneration and colony stimulating factor 1 receptor (CSF1R) processing in microglia.
iScience (2021-12-09)
Giulio Morozzi, Julian Rothen, Gauthier Toussaint, Katrina De Lange, Katrin Westritschnig, Arno Doelemeyer, Vanessa Pitiot Ueberschlag, Peter Kahle, Christian Lambert, Michael Obrecht, Nicolau Beckmann, Veronique Ritter, Moh Panesar, Daniela Stauffer, Isabelle Garnier, Matthias Mueller, Danilo Guerini, Caroline Gubser Keller, Judith Knehr, Guglielmo Roma, Michael Bidinosti, Sophie Brachat, Frederic Morvan, Mara Fornaro
ABSTRACT
Inflammatory responses are crucial for regeneration following peripheral nerve injury (PNI). PNI triggers inflammatory responses at the site of injury. The DNA-sensing receptor cyclic GMP-AMP synthase (cGAS) and its downstream effector stimulator of interferon genes (STING) sense foreign and self-DNA and trigger type I interferon (IFN) immune responses. We demonstrate here that following PNI, the cGAS/STING pathway is upregulated in the sciatic nerve of naive rats and dysregulated in old rats. In a nerve crush mouse model where STING is knocked out, myelin content in sciatic nerve is increased resulting in accelerated functional axon recovery. STING KO mice have lower macrophage number in sciatic nerve and decreased microglia activation in spinal cord 1 week post injury. STING activation regulated processing of colony stimulating factor 1 receptor (CSF1R) and microglia survival in vitro. Taking together, these data highlight a previously unrecognized role of STING in the regulation of nerve regeneration.
Sigma-Aldrich
Anti-Neurofilament 200kDa Antibody, clone NE14, Alexa Fluor 555 Conjugate, clone NE14, from mouse, ALEXA FLUOR 555
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture