Merck
  • Designed Ankyrin Repeat Proteins as Novel Binders for Ultrasound Molecular Imaging.
Designed Ankyrin Repeat Proteins as Novel Binders for Ultrasound Molecular Imaging.
Ultrasound in medicine & biology (2021-06-20)
Alexandra Kosareva, Mukesh Punjabi, Amanda Ochoa-Espinosa, Lifen Xu, Jonas V Schaefer, Birgit Dreier, Andreas Plückthun, Beat A Kaufmann
ABSTRACT
Clinical translation of ultrasound molecular imaging will depend on the development of binders that can easily be generated, manufactured and coupled, and that are compatible with in vivo use. We describe targeted microbubbles (MBs) using designed ankyrin repeat proteins (DARPins) as a novel class of such translatable binders. Candidate DARPin binders for vascular cell adhesion molecule 1, an endothelial cell adhesion molecule involved in inflammatory processes, were selected using ribosome display and coupled to MBs. Flow-chamber assays of five MBs carrying high-affinity binders showed selective retention on endothelial cells activated by tumor necrosis factor-α for two binders compared with a MB carrying a control DARPin. In vivo ultrasound molecular imaging in a murine hind-limb inflammation model demonstrated up to a fourfold signal enhancement for three of the five MBs versus control. However, there was no correlation between results from flow-chamber assays and in vivo imaging. Thus, we conclude that ultrasound molecular imaging of inflammation using DARPin binders is feasible per se, but that screening of candidates cannot be accomplished with flow-chamber assays as used in our study.