Merck
  • Design, Synthesis, and In Vitro Antiproliferative Activity of Hydantoin and Purine Derivatives with the 4-Acetylphenylpiperazinylalkyl Moiety.
Design, Synthesis, and In Vitro Antiproliferative Activity of Hydantoin and Purine Derivatives with the 4-Acetylphenylpiperazinylalkyl Moiety.
Materials (Basel, Switzerland) (2021-08-08)
Agnieszka Zagórska, Anna Czopek, Anna Jaromin, Magdalena Mielczarek-Puta, Marta Struga, Dorota Stary, Marek Bajda
ABSTRACT
Cancer represents one of the most serious health problems and the second leading cause of death around the world. Heterocycles, due to their prevalence in nature as well as their structural and chemical diversity, play an immensely important role in anti-cancer drug discovery. In this paper, a series of hydantoin and purine derivatives containing a 4-acetylphenylpiperazinylalkyl moiety were designed, synthesized, and biologically evaluated for their anticancer activity on selected cancer cell lines (PC3, SW480, SW620). Compound 4, a derivative of 3',4'-dihydro-2'H-spiro[imidazolidine-4,1'-naphthalene]-2,5-dione, was the most effective against SW480, SW620, and PC3 cancer cell lines. Moreover, 4 has high tumor-targeting selectivity. Based on docking studies, it was concluded that R isomers of 3',4'-dihydro-2'H-spiro[imidazolidine-4,1'-naphthalene]-2,5-dione could be further studied as promising scaffolds for the development of thymidine phosphorylase inhibitors.
Sigma-Aldrich
Thymidine Phosphorylase, recombinant from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution, ≥900 units/mL, 0.2 μm filtered
Pricing and availability is not currently available.