Merck
Get up to 22% off for Pi Day until 3/26.Save Now
  • Biocytin wide-field bipolar cells in rabbit retina selectively contact blue cones.
Biocytin wide-field bipolar cells in rabbit retina selectively contact blue cones.
The Journal of comparative neurology (2007-11-09)
Margaret A MacNeil, Paulette A Gaul
ABSTRACT
The biocytin wide-field bipolar cell in rabbit retina has a broad axonal arbor in layer 5 of the inner plexiform layer and a wide dendritic arbor that does not contact all cones in its dendritic field. The purpose of our study was to identify the types of cones that this cell contacts. We identified the bipolar cells by selective uptake of biocytin, labeled the cones with peanut agglutinin, and then used antibodies against blue cone opsin and red-green cone opsin to identify the individual cone types. The biocytin-labeled cells selectively contacted cones whose outer segments stained for blue cone opsin and avoided cones that did not. We conclude that the biocytin wide-field bipolar cell is an ON blue cone bipolar cell in the rabbit retina and is homologous to the blue cone bipolar cells that have been previously described in primate, mouse, and ground squirrel retinas.
Sigma-Aldrich
Biocytin hydrochloride, ≥98% (TLC)
Sigma-Aldrich
Biocytin, ≥98% (TLC)