Merck
  • The evaluation of the role of beta-hydroxy fatty acids on chronic inflammation and insulin resistance.
The evaluation of the role of beta-hydroxy fatty acids on chronic inflammation and insulin resistance.
Mediators of inflammation (2007-03-30)
A S Soydan, H S Dokmetas, M Cetin, A Koyuncu, E Kaptanoglu, H Elden
ABSTRACT
Beta-hydroxy fatty acids are a major component of lipid A moiety of lipopolysaccharide. We aimed to investigate the role of free beta-hydroxy fatty acids on inflammation, as well as to evaluate their effects on cytokine release from human blood cells, and whether they exist in plasma of patients with chronic inflammatory diseases with/without insulin resistance. Peripheral venous blood was incubated with beta-hydroxy lauric and beta-hydroxy myristic acids (each 100 ng, 1 microg, 10 microg/mL) up to 24 hours. Cytokines were measured from culture media and plasma. Free fatty acids and biochemical parameters were also measured from patients' plasma. Only beta-hydroxy lauric acid significantly stimulated interleukin-6 production at 10 microg/mL compared to control (533.9 +/- 218.1 versus 438.3 +/- 219.6 pg/mL, P < .05). However, free beta-hydroxy lauric and myristic acids were not found in patients' plasma. Therefore, free beta-hydroxy lauric and myristic acids do not seem to have a role on sterile inflammation in chronic inflammatory diseases associated with insulin resistance.
Sigma-Aldrich
DL-β-Hydroxylauric acid, ≥99% (GC)
Sigma-Aldrich
DL-β-Hydroxymyristic acid, ≥98%