Merck
  • Quantification of Bacteria Residing in Caenorhabditis elegans Intestine.
Quantification of Bacteria Residing in Caenorhabditis elegans Intestine.
Bio-protocol (2021-03-05)
M Fernanda Palominos, Andrea Calixto
ABSTRACT
Quantification of intestinal colonization by pathogenic or commensal bacteria constitute a critical part of the analysis to understand host-microbe interactions during different time points of their interplay. Here we detail a method to isolate non-pathogenic and pathogenic bacteria from C. elegans intestines, and classify gut phenotypes induced by bacterial pathogens using fluorescently-tagged bacteria. Furthermore, these methods can be used to isolate and identify new culturable bacterial species from natural microbiomes of wild nematodes.
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Gentamicin sulfate, meets EP, USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
USP
Levamisole hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, ≥99.5%